Assay Details
| Gene Symbol: LOC4332563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775955.2 | XP_015631441.1 | 8 - 9 | 981 | 108 | |
| GenBank mRNA | AK069075.1 | - | 8 - 9 | 941 | 108 | |
| AY338235.1 | - | 8 - 9 | 893 | 108 | ||
More Information
Set Membership: |
|

LOC4332563
Genomic Map