Assay Details
| Gene Symbol: LOC9272301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773585.2 | XP_015629071.1 | 7 - 8 | 1049 | 85 | |
| GenBank mRNA | AK241138.1 | - | 7 - 8 | 955 | 85 | |
More Information
Set Membership: |
|

LOC9272301
Genomic Map