Assay Details
| Gene Symbol: LOC9272301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773585.2 | XP_015629071.1 | 9 - 10 | 1514 | 84 | |
| GenBank mRNA | AK241138.1 | - | 9 - 10 | 1420 | 84 | |
More Information
Set Membership: |
|

LOC9272301
Genomic Map