Assay Details
| Gene Symbol: LOC4332774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777653.2 | XP_015633139.1 | 7 - 8 | 909 | 236 | |
| GenBank mRNA | AK243527.1 | - | 7 - 8 | 912 | 236 | |
More Information
Set Membership: |
|

LOC4332774
Genomic Map