Assay Details
| Gene Symbol: LOC4332822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775133.2 | XP_015630619.1 | 14 - 15 | 1483 | 69 | |
| GenBank mRNA | AK058969.1 | - | 1 - 2 | 140 | 69 | |
| AK102852.1 | - | 14 - 15 | 1444 | 69 | ||
| AY017415.1 | - | 14 - 15 | 1460 | 69 | ||
More Information
Set Membership: |
|

LOC4332822
Genomic Map