Assay Details
| Gene Symbol: LOC4332822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775133.2 | XP_015630619.1 | 17 - 18 | 2467 | 101 | |
| GenBank mRNA | AK058460.1 | - | 3 - 4 | 615 | 101 | |
| AK058969.1 | - | 4 - 5 | 1124 | 101 | ||
| AK102852.1 | - | 17 - 18 | 2427 | 101 | ||
| AY017415.1 | - | 17 - 18 | 2444 | 101 | ||
More Information
Set Membership: |
|

LOC4332822
Genomic Map