Assay Details
| Gene Symbol: LOC4332822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775133.2 | XP_015630619.1 | 5 - 6 | 439 | 128 | |
| GenBank mRNA | AK102852.1 | - | 5 - 6 | 400 | 128 | |
| AY017415.1 | - | 5 - 6 | 416 | 128 | ||
More Information
Set Membership: |
|

LOC4332822
Genomic Map