Assay Details
| Gene Symbol: LOC4333006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773540.2 | XP_015629026.1 | 1 - 2 | 432 | 92 | |
| GenBank mRNA | AK059680.1 | - | 1 - 2 | 416 | 92 | |
| AK099133.1 | - | 1 - 2 | 389 | 92 | ||
More Information
Set Membership: |
|

LOC4333006
Genomic Map