Assay Details
| Gene Symbol: LOC4333070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774598.2 | XP_015630084.1 | 4 - 5 | 712 | 121 | |
| XM_015774599.2 | XP_015630085.1 | 4 - 5 | 763 | 121 | ||
| GenBank mRNA | AK099537.1 | - | 4 - 5 | 765 | 121 | |
More Information
Set Membership: |
|

LOC4333070
Genomic Map