Assay Details
| Gene Symbol: LOC4333431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776969.2 | XP_015632455.1 | 3 - 4 | 682 | 90 | |
| GenBank mRNA | AK065225.1 | - | 2 - 3 | 1339 | 90 | |
| AK065981.1 | - | 3 - 4 | 646 | 90 | ||
More Information
Set Membership: |
|

LOC4333431
Genomic Map