Assay Details
| Gene Symbol: LOC4333468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776748.2 | XP_015632234.1 | 1 - 2 | 158 | 137 | |
| GenBank mRNA | AK071474.1 | - | 1 - 2 | 93 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776748.2 transcripts.
Set Membership: |
|

LOC4333468
Genomic Map