Assay Details
| Gene Symbol: LOC4333540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776880.2 | XP_015632366.1 | 5 - 6 | 1227 | 66 | |
| GenBank mRNA | AK059114.1 | - | 5 - 6 | 1223 | 66 | |
More Information
Set Membership: |
|

LOC4333540
Genomic Map