Assay Details
| Gene Symbol: LOC4333634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772938.2 | XP_015628424.1 | 1 | 1042 | 67 | |
| GenBank mRNA | AK058637.1 | - | 1 | 1039 | 67 | |
| GQ848070.1 | - | 1 | 834 | 67 | ||
More Information
Set Membership: |
|

LOC4333634
Genomic Map