Assay Details
| Gene Symbol: LOC4333726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775514.2 | XP_015631000.1 | 2 - 3 | 599 | 113 | |
| GenBank mRNA | AK067939.1 | - | 2 - 3 | 277 | 113 | |
| AK069994.1 | - | 2 - 3 | 276 | 113 | ||
| AK070543.1 | - | 2 - 3 | 600 | 112 | ||
| FJ940209.1 | - | 2 - 3 | 219 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775514.2 transcripts.
Set Membership: |
|

LOC4333726
Genomic Map