Assay Details
| Gene Symbol: LOC4333726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775514.2 | XP_015631000.1 | 4 - 5 | 1931 | 80 | |
| GenBank mRNA | AJ557442.1 | - | 2 - 3 | 1226 | 80 | |
| AK061393.1 | - | 1 - 2 | 93 | 80 | ||
| AK067939.1 | - | 4 - 5 | 1609 | 80 | ||
| AK069994.1 | - | 4 - 5 | 1608 | 80 | ||
| AK104713.1 | - | 1 - 2 | 93 | 80 | ||
| FJ940209.1 | - | 4 - 5 | 1551 | 80 | ||
More Information
Set Membership: |
|

LOC4333726
Genomic Map