Assay Details
| Gene Symbol: LOC9267831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB067652.1 | - | 4 - 5 | 1012 | 106 | |
| AK070804.1 | - | 4 - 5 | 1158 | 106 | ||
| AK072691.1 | - | 5 - 6 | 1490 | 106 | ||
| AK099012.1 | - | 4 - 5 | 1133 | 106 | ||
More Information
Set Membership: |
|

LOC9267831
Genomic Map