Assay Details
| Gene Symbol: LOC4333941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776796.2 | XP_015632282.1 | 4 - 5 | 2140 | 154 | |
| XM_015776797.2 | XP_015632283.1 | 4 - 5 | 2137 | 154 | ||
| GenBank mRNA | AK058989.1 | - | 2 - 3 | 706 | 154 | |
More Information
Set Membership: |
|

LOC4333941
Genomic Map