Assay Details
| Gene Symbol: LOC4334069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775958.2 | XP_015631444.1 | 2 - 3 | 722 | 74 | |
| GenBank mRNA | AK059838.1 | - | 2 - 3 | 672 | 74 | |
| AK100790.1 | - | 2 - 3 | 75 | |||
| BT131836.1 | - | 1 - 2 | 70 | 74 | ||
More Information
Set Membership: |
|

LOC4334069
Genomic Map