Assay Details
| Gene Symbol: LOC4334069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775958.2 | XP_015631444.1 | 3 - 4 | 820 | 72 | |
| GenBank mRNA | AK059838.1 | - | 3 - 4 | 770 | 72 | |
| AK062852.1 | - | 1 - 2 | 38 | 72 | ||
| AK100790.1 | - | 3 - 4 | 775 | 72 | ||
| BT131836.1 | - | 2 - 3 | 168 | 72 | ||
More Information
Set Membership: |
|

LOC4334069
Genomic Map