Assay Details
| Gene Symbol: LOC4334069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775958.2 | XP_015631444.1 | 4 - 5 | 882 | 111 | |
| GenBank mRNA | AK059838.1 | - | 4 - 5 | 832 | 111 | |
| AK062852.1 | - | 2 - 3 | 100 | 111 | ||
| AK100790.1 | - | 4 - 5 | 837 | 111 | ||
| BT131836.1 | - | 3 - 4 | 230 | 111 | ||
More Information
Set Membership: |
|

LOC4334069
Genomic Map