Assay Details
| Gene Symbol: LOC4334089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772606.2 | XP_015628092.1 | 11 - 12 | 1486 | 98 | |
| GenBank mRNA | AK101069.1 | - | 11 - 12 | 1465 | 98 | |
| GQ982584.1 | - | 11 - 12 | 1271 | 98 | ||
More Information
Set Membership: |
|

LOC4334089
Genomic Map