Assay Details
| Gene Symbol: LOC4334114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775683.2 | XP_015631169.1 | 3 - 4 | 980 | 99 | |
| XM_015775684.2 | XP_015631170.1 | 3 - 4 | 617 | 99 | ||
| GenBank mRNA | AK065044.1 | - | 2 - 3 | 1395 | 99 | |
| AK122100.1 | - | 2 - 3 | 445 | 99 | ||
More Information
Set Membership: |
|

LOC4334114
Genomic Map