Assay Details
| Gene Symbol: LOC4334269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776995.2 | XP_015632481.1 | 8 - 9 | 2200 | 102 | |
| XM_015776996.2 | XP_015632482.1 | 10 - 11 | 1720 | 102 | ||
| GenBank mRNA | AK065113.1 | - | 8 - 9 | 1766 | 102 | |
More Information
Set Membership: |
|

LOC4334269
Genomic Map