Assay Details
| Gene Symbol: LOC4334272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772721.2 | XP_015628207.1 | 12 - 13 | 1921 | 70 | |
| XM_015772722.2 | XP_015628208.1 | 12 - 13 | 1921 | 70 | ||
| GenBank mRNA | AK059632.1 | - | 1 - 2 | 77 | 70 | |
| AK103310.1 | - | 12 - 13 | 1371 | 70 | ||
More Information
Set Membership: |
|

LOC4334272
Genomic Map