Assay Details
| Gene Symbol: LOC4334272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772721.2 | XP_015628207.1 | 5 - 6 | 1041 | 149 | |
| XM_015772722.2 | XP_015628208.1 | 5 - 6 | 1041 | 149 | ||
| GenBank mRNA | AK103310.1 | - | 5 - 6 | 491 | 149 | |
More Information
Set Membership: |
|

LOC4334272
Genomic Map