Assay Details
| Gene Symbol: LOC4334272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772721.2 | XP_015628207.1 | 9 - 10 | 1471 | 85 | |
| XM_015772722.2 | XP_015628208.1 | 9 - 10 | 1471 | 85 | ||
| GenBank mRNA | AK103310.1 | - | 9 - 10 | 921 | 85 | |
More Information
Set Membership: |
|

LOC4334272
Genomic Map