Assay Details
| Gene Symbol: LOC4334513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775065.2 | XP_015630551.1 | 3 - 4 | 644 | 74 | |
| GenBank mRNA | AK060962.1 | - | 3 - 4 | 652 | 74 | |
| AK070287.1 | - | 3 - 4 | 550 | 74 | ||
| KX434728.1 | - | 3 - 4 | 399 | 74 | ||
More Information
Set Membership: |
|

LOC4334513
Genomic Map