Assay Details
| Gene Symbol: LOC4334571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773883.2 | XP_015629369.1 | 1 - 2 | 367 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334571| Gene Symbol: LOC4334571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773883.2 | XP_015629369.1 | 1 - 2 | 367 | 64 | |
Set Membership: |
|