Assay Details
| Gene Symbol: LOC4334698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774276.1 | XP_015629762.1 | 5 - 6 | 1309 | 128 | |
| GenBank mRNA | AK061773.1 | - | 5 - 6 | 980 | 128 | |
| AK098986.1 | - | 5 - 6 | 968 | 128 | ||
| AK105896.1 | - | 5 - 6 | 980 | 128 | ||
More Information
Set Membership: |
|

LOC4334698
Genomic Map