Assay Details
| Gene Symbol: LOC4334700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774271.2 | XP_015629757.1 | 12 - 13 | 1273 | 59 | |
| XM_015774273.2 | XP_015629759.1 | 12 - 13 | 1274 | 59 | ||
| GenBank mRNA | AK070689.1 | - | 6 - 7 | 574 | 59 | |
| AK102903.1 | - | 12 - 13 | 1211 | 59 | ||
More Information
Set Membership: |
|

LOC4334700
Genomic Map