Assay Details
| Gene Symbol: LOC4334821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777520.2 | XP_015633006.1 | 1 - 2 | 252 | 103 | |
| XM_015777521.2 | XP_015633007.1 | 1 - 2 | 250 | 103 | ||
| GenBank mRNA | AK064339.1 | - | 1 - 2 | 190 | 103 | |
| AK103879.1 | - | 1 - 2 | 180 | 103 | ||
More Information
Set Membership: |
|

LOC4334821
Genomic Map