Assay Details
Gene Symbol: LOC4334821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777521.2 | XP_015633007.1 | 7 - 8 | 1089 | 77 | |
GenBank mRNA | AK064339.1 | - | 7 - 8 | 1029 | 77 | |
AK103879.1 | - | 7 - 8 | 1019 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
