Assay Details
| Gene Symbol: LOC4334829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775446.2 | XP_015630932.1 | 3 - 4 | 1807 | 154 | |
| XM_015775447.2 | XP_015630933.1 | 3 - 4 | 2939 | 154 | ||
| XM_015775448.2 | XP_015630934.1 | 4 - 5 | 3012 | 154 | ||
| XM_015775449.2 | XP_015630935.1 | 4 - 5 | 1855 | 154 | ||
| GenBank mRNA | AK060748.1 | - | 5 - 6 | 1792 | 154 | |
| AK072499.1 | - | 2 - 3 | 3226 | 154 | ||
More Information
Set Membership: |
|

LOC4334829
Genomic Map