Assay Details
| Gene Symbol: LOC4334829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775446.2 | XP_015630932.1 | 6 - 7 | 2153 | 76 | |
| XM_015775447.2 | XP_015630933.1 | 6 - 7 | 3285 | 76 | ||
| XM_015775448.2 | XP_015630934.1 | 7 - 8 | 3358 | 76 | ||
| XM_015775449.2 | XP_015630935.1 | 7 - 8 | 2201 | 76 | ||
| GenBank mRNA | AK060748.1 | - | 8 - 9 | 2138 | 76 | |
More Information
Set Membership: |
|

LOC4334829
Genomic Map