Assay Details
| Gene Symbol: LOC4334857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776770.2 | XP_015632256.1 | 1 - 2 | 323 | 58 | |
| GenBank mRNA | AK105297.1 | - | 1 - 2 | 255 | 58 | |
More Information
Set Membership: |
|

LOC4334857
Genomic Map