Assay Details
| Gene Symbol: LOC4334857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776770.2 | XP_015632256.1 | 4 - 5 | 979 | 129 | |
| XM_015776771.2 | XP_015632257.1 | 4 - 5 | 975 | 129 | ||
| GenBank mRNA | AK073135.1 | - | 4 - 5 | 922 | 129 | |
| AK105297.1 | - | 4 - 5 | 911 | 129 | ||
More Information
Set Membership: |
|

LOC4334857
Genomic Map