Assay Details
| Gene Symbol: LOC4334917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778347.2 | XP_015633833.1 | 1 - 2 | 61 | 83 | |
| GenBank mRNA | AK058573.1 | - | 1 - 2 | 61 | 83 | |
| FJ746894.1 | - | 1 - 2 | 61 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015778347.2 transcripts.
Set Membership: |
|

LOC4334917
Genomic Map