Assay Details
| Gene Symbol: LOC4334929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779970.2 | XP_015635456.1 | 1 | 2463 | 69 | |
| GenBank mRNA | AK073722.1 | - | 1 | 2453 | 69 | |
| AK103144.1 | - | 1 | 2453 | 69 | ||
More Information
Set Membership: |
|

LOC4334929
Genomic Map