Assay Details
| Gene Symbol: LOC4336013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781062.2 | XP_015636548.1 | 3 - 4 | 1382 | 81 | |
| GenBank mRNA | AK121306.1 | - | 3 - 4 | 1308 | 81 | |
More Information
Set Membership: |
|

LOC4336013
Genomic Map