Assay Details
| Gene Symbol: LOC4336193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781531.2 | XP_015637017.1 | 3 - 4 | 1178 | 118 | |
| XM_015781532.2 | XP_015637018.1 | 3 - 4 | 1206 | 118 | ||
| GenBank mRNA | AK064143.1 | - | 3 - 4 | 1032 | 118 | |
More Information
Set Membership: |
|

LOC4336193
Genomic Map