Assay Details
| Gene Symbol: LOC4336389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781208.1 | XP_015636694.1 | 5 - 6 | 594 | 73 | |
| XM_015781209.1 | XP_015636695.1 | 5 - 6 | 630 | 73 | ||
| XM_015781210.1 | XP_015636696.1 | 5 - 6 | 594 | 73 | ||
| GenBank mRNA | AK066850.1 | - | 3 - 4 | 1714 | 73 | |
| AK068772.1 | - | 5 - 6 | 490 | 73 | ||
More Information
Set Membership: |
|

LOC4336389
Genomic Map