Assay Details
| Gene Symbol: LOC4336400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778749.2 | XP_015634235.1 | 2 - 3 | 778 | 58 | |
| GenBank mRNA | AK062955.1 | - | 2 - 3 | 603 | 58 | |
| KY050768.1 | - | 2 - 3 | 432 | 58 | ||
More Information
Set Membership: |
|

LOC4336400
Genomic Map