Assay Details
| Gene Symbol: LOC4336691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778951.2 | XP_015634437.1 | 4 - 5 | 782 | 74 | |
| GenBank mRNA | AK101849.1 | - | 4 - 5 | 663 | 74 | |
More Information
Set Membership: |
|

LOC4336691
Genomic Map