Assay Details
| Gene Symbol: LOC4336788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778626.2 | XP_015634112.1 | 2 - 3 | 348 | 114 | |
| GenBank mRNA | AK105286.1 | - | 2 - 3 | 324 | 114 | |
More Information
Set Membership: |
|

LOC4336788
Genomic Map