Assay Details
| Gene Symbol: LOC4336875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778145.2 | XP_015633631.1 | 2 - 3 | 1134 | 57 | |
| GenBank mRNA | AB004865.1 | - | 2 - 3 | 1083 | 57 | |
| AK107376.1 | - | 2 - 3 | 1098 | 57 | ||
More Information
Set Membership: |
|

LOC4336875
Genomic Map