Assay Details
| Gene Symbol: LOC4337307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779446.2 | XP_015634932.1 | 1 - 2 | 594 | 84 | |
| GenBank mRNA | AK105321.1 | - | 1 - 2 | 98 | 84 | |
More Information
Set Membership: |
|

LOC4337307
Genomic Map