Assay Details
| Gene Symbol: LOC4337511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780693.2 | XP_015636179.2 | 2 - 3 | 371 | 71 | |
| GenBank mRNA | AK102150.1 | - | 2 - 3 | 324 | 71 | |
More Information
Set Membership: |
|

LOC4337511
Genomic Map