Assay Details
| Gene Symbol: LOC4337561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782139.2 | XP_015637625.2 | 1 - 2 | 255 | 75 | |
| XM_015782140.2 | XP_015637626.2 | 1 - 2 | 254 | 75 | ||
| GenBank mRNA | AK063761.1 | - | 1 - 2 | 229 | 75 | |
More Information
Set Membership: |
|

LOC4337561
Genomic Map