Assay Details
| Gene Symbol: LOC4337561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782139.2 | XP_015637625.2 | 5 - 6 | 637 | 71 | |
| GenBank mRNA | AK063761.1 | - | 5 - 6 | 611 | 71 | |
More Information
Set Membership: |
|

LOC4337561
Genomic Map