Assay Details
| Gene Symbol: LOC4337561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782139.2 | XP_015637625.2 | 6 - 7 | 856 | 93 | |
| XM_015782140.2 | XP_015637626.2 | 6 - 7 | 852 | 93 | ||
| GenBank mRNA | AK063761.1 | - | 6 - 7 | 830 | 93 | |
More Information
Set Membership: |
|

LOC4337561
Genomic Map